Learn More
Medchemexpress LLC Fc gamma RIIA/CD32a 100ug
SDP
Catalog No. 5000185081
Encompass_Preferred
Recombinant human Fc gamma RIIA/CD32a (His-tag) is a HEK293-expressed, lyophilized receptor protein designed for in vitro studies of IgG-Fc interactions, antibody characterization, and receptor biology. The protein is provided with low endotoxin and validated binding activity for biosensor and ELISA formats.
  • Expressed in HEK293 cells for native human-like post-translational modifications.
  • His tag for straightforward capture and detection.
  • Lyophilized formulation for stable storage and shipping.
  • Low endotoxin level (<1 EU/μg) suitable for many bioassays.
  • Validated binding activity (BLI KD 0.98 μM; ED50 3.612 μg/mL).
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-85081 Supplier Medchemexpress LLC Supplier No. HYP70711100UG
Recombinant human Fc gamma RIIA/CD32a (His-tag) is a HEK293-expressed, lyophilized receptor protein designed for in vitro studies of IgG-Fc interactions, antibody characterization, and receptor biology. The protein is provided with low endotoxin and validated binding activity for biosensor and ELISA formats.
  • Expressed in HEK293 cells for native human-like post-translational modifications.
  • His tag for straightforward capture and detection.
  • Lyophilized formulation for stable storage and shipping.
  • Low endotoxin level (<1 EU/μg) suitable for many bioassays.
  • Validated binding activity (BLI KD 0.98 μM; ED50 3.612 μg/mL).
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw cycles.