Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems CHO/Human CD32a (131H) Stable Cell Line, Low Expression, 2 Vials (1 mL, ≥5x10^6 cells/vial)

Catalog No. 502370256
Encompass_Preferred

Stable CHO cell line expressing full-length human Fc gamma RIIa (CD32a) 131H allotype at low expression. Supplied frozen in cryopreserved vials, this cell line is designed for cell-based CD32a binding and crosslinking assays and for evaluating CD32a-dependent antibody agonism.

  • Expresses full-length human CD32a (131H) allotype.
  • Low expression level suitable for sensitivity testing.
  • Supplied as two cryopreserved vials; 1 mL per vial.
  • Each vial contains ≥ 5x10^6 cells.
  • Host cell: CHO; adherent growth properties.
  • Selection marker: hygromycin B (20 μg/mL).
  • Recommended growth medium: F-12K with 10% FBS.
  • Storage: keep frozen in liquid nitrogen.
  • Quality tested: mycoplasma negative and sterility tested.

Catalog No. 50-237-0256 Supplier ACROBiosystems Supplier No. SCCHOATP061L2VIALS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stable CHO cell line expressing full-length human Fc gamma RIIa (CD32a) 131H allotype at low expression. Supplied frozen in cryopreserved vials, this cell line is designed for cell-based CD32a binding and crosslinking assays and for evaluating CD32a-dependent antibody agonism.

  • Expresses full-length human CD32a (131H) allotype.
  • Low expression level suitable for sensitivity testing.
  • Supplied as two cryopreserved vials; 1 mL per vial.
  • Each vial contains ≥ 5x10^6 cells.
  • Host cell: CHO; adherent growth properties.
  • Selection marker: hygromycin B (20 μg/mL).
  • Recommended growth medium: F-12K with 10% FBS.
  • Storage: keep frozen in liquid nitrogen.
  • Quality tested: mycoplasma negative and sterility tested.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.