Learn More
Medchemexpress LLC Fc gamma RIIA/CD32a 500ug
SDP
Catalog No. 5000184484
Encompass_Preferred
Recombinant human Fc gamma RIIA (CD32a) extracellular domain (H167R allotype), expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized, the 183 amino-acid protein is provided with low endotoxin and a buffered formulation to support stability; it is suitable for binding studies, surface plasmon resonance, and other biochemical assays when reconstituted and handled according to storage recommendations.
  • Recombinant extracellular domain (183 a.a.) for receptor binding studies.
  • HEK293 expression ensures mammalian post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Low endotoxin (<1 EU/μg) for sensitive bioassays.
  • Lyophilized formulation for extended shelf life and transport.
  • Stable under recommended storage and aliquoting conditions.
Catalog No. 50-001-84484 Supplier Medchemexpress LLC Supplier No. HYP70478500UG
Recombinant human Fc gamma RIIA (CD32a) extracellular domain (H167R allotype), expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized, the 183 amino-acid protein is provided with low endotoxin and a buffered formulation to support stability; it is suitable for binding studies, surface plasmon resonance, and other biochemical assays when reconstituted and handled according to storage recommendations.
  • Recombinant extracellular domain (183 a.a.) for receptor binding studies.
  • HEK293 expression ensures mammalian post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Low endotoxin (<1 EU/μg) for sensitive bioassays.
  • Lyophilized formulation for extended shelf life and transport.
  • Stable under recommended storage and aliquoting conditions.