Learn More
Medchemexpress LLC Fc gamma RIIA/CD32a 50ug | 50ug
SDP
Catalog No. 5000193581
Encompass_Preferred
Fc gamma RIIA/CD32a protein belongs to the FC family of receptor proteins and is a type 1 transmembrane glycoprotein that binds immunoglobulin subtypes IgG1-4 and can bind c-reactive protein (CRP) with high affinity It plays an important role in immune regulation in many infections including malaria Fc gamma RIIA/CD32a Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIIA/CD32a protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93581 Supplier Medchemexpress LLC Supplier No. HYP7826550UG
Fc gamma RIIA/CD32a protein belongs to the FC family of receptor proteins and is a type 1 transmembrane glycoprotein that binds immunoglobulin subtypes IgG1-4 and can bind c-reactive protein (CRP) with high affinity It plays an important role in immune regulation in many infections including malaria Fc gamma RIIA/CD32a Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIIA/CD32a protein expressed by HEK293 with C-His labeled tag