Learn More
Medchemexpress LLC Fc gamma RIIA/CD32a 5ug
SDP
Catalog No. 5000184821
Encompass_Preferred
Recombinant human FcγRIIA/CD32a ectodomain (183 amino acids, H167R) expressed in HEK293 cells and supplied lyophilized in 20 mM PB, 150 mM NaCl, pH 7.4. The protein is intended for in vitro binding and receptor-ligand interaction studies; reconstituted material should be aliquoted and stored frozen to preserve activity.
  • Expressed in HEK293 cells to preserve native human glycosylation patterns.
  • C-terminal His tag enables straightforward affinity purification and detection.
  • 183 amino acid ectodomain suitable for receptor-ligand interaction assays.
  • Lyophilized formulation for extended shelf life and convenient transport.
  • Buffered in physiological saline for compatibility with common assay buffers.
  • Stable frozen storage with recommended aliquoting to minimize freeze-thaw cycles.
Catalog No. 50-001-84821 Supplier Medchemexpress LLC Supplier No. HYP704785UG
Recombinant human FcγRIIA/CD32a ectodomain (183 amino acids, H167R) expressed in HEK293 cells and supplied lyophilized in 20 mM PB, 150 mM NaCl, pH 7.4. The protein is intended for in vitro binding and receptor-ligand interaction studies; reconstituted material should be aliquoted and stored frozen to preserve activity.
  • Expressed in HEK293 cells to preserve native human glycosylation patterns.
  • C-terminal His tag enables straightforward affinity purification and detection.
  • 183 amino acid ectodomain suitable for receptor-ligand interaction assays.
  • Lyophilized formulation for extended shelf life and convenient transport.
  • Buffered in physiological saline for compatibility with common assay buffers.
  • Stable frozen storage with recommended aliquoting to minimize freeze-thaw cycles.