Learn More
Genscript Corporation Fc gamma RIIA/CD32a H167 Hi
Supplier: Genscript Corporation Z04250500
The Fc gamma Rs have been divided into three classes based on close relationships in their extracellular domains these groups are designated Fc gamma RI (also known as CD64) Fc gamma RII (CD32) and Fc gamma RIII (CD16) Each group may be encoded by multiple genes and exist in different isoforms depending on species and cell type