Learn More
Medchemexpress LLC Fc gamma RIIA (CD32a) recombinant human Fc-fusion protein, H167R | 9000-83-3 | 50 UG
SDP
Catalog No. 5000292625
Encompass_Preferred
Recombinant human Fc gamma RIIA (CD32a) Fc-fusion protein, H167R variant, produced in HEK293 cells. Intended for biochemical and cell-based studies of antibody Fc interactions and receptor binding; provided as a recombinant extracellular domain fused to a human Fc to improve stability and detection.
  • Recombinant human extracellular domain with H167R mutation for functional assays.
  • HEK293 expression ensures mammalian glycosylation and proper folding.
  • C-terminal human Fc fusion increases stability and simplifies detection.
  • Suitable for receptor binding, SPR, ELISA, and cell-based assays.
  • Provided in a research-grade recombinant protein format for in vitro studies.
Catalog No. 50-002-92625 Supplier Medchemexpress LLC Supplier No. HYP7564450UG
Recombinant human Fc gamma RIIA (CD32a) Fc-fusion protein, H167R variant, produced in HEK293 cells. Intended for biochemical and cell-based studies of antibody Fc interactions and receptor binding; provided as a recombinant extracellular domain fused to a human Fc to improve stability and detection.
  • Recombinant human extracellular domain with H167R mutation for functional assays.
  • HEK293 expression ensures mammalian glycosylation and proper folding.
  • C-terminal human Fc fusion increases stability and simplifies detection.
  • Suitable for receptor binding, SPR, ELISA, and cell-based assays.
  • Provided in a research-grade recombinant protein format for in vitro studies.