Learn More
Medchemexpress LLC Fc gamma RIIA (CD32a) recombinant protein | ≥95.0% | 500 UG
SDP
Catalog No. 5000292446
Encompass_Preferred
Recombinant His-tagged Fc gamma RIIA (CD32a) ectodomain from rhesus macaque, expressed in HEK293 cells and supplied as a lyophilized powder. It is intended for in vitro binding studies, immunoassays, and biophysical characterization, with controlled endotoxin and purity specifications.
Catalog No. 50-002-92446 Supplier Medchemexpress LLC Supplier No. HYP75647500UG
Recombinant His-tagged Fc gamma RIIA (CD32a) ectodomain from rhesus macaque, expressed in HEK293 cells and supplied as a lyophilized powder. It is intended for in vitro binding studies, immunoassays, and biophysical characterization, with controlled endotoxin and purity specifications.