Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fc gamma RIIB/CD32b 100ug
SDP

Catalog No. 5000184490
Encompass_Preferred

Recombinant human Fc gamma RIIB (CD32b) extracellular domain produced in HEK293 cells and supplied as a His-tagged, lyophilized protein. Purified to >95% by reducing SDS-PAGE, the protein is formulated for stability and is intended for biochemical and biophysical studies of IgG-receptor interactions, assay development, and receptor characterization.

  • Recombinant extracellular domain expressed in HEK293 cells.
  • C-terminal 6xHis tag for straightforward detection and capture.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Molecular weight approximately 25-30 kDa.
  • Suitable for binding studies, SPR/BLI, and assay development.

Catalog No. 50-001-84490 Supplier Medchemexpress LLC Supplier No. HYP70491100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Fc gamma RIIB (CD32b) extracellular domain produced in HEK293 cells and supplied as a His-tagged, lyophilized protein. Purified to >95% by reducing SDS-PAGE, the protein is formulated for stability and is intended for biochemical and biophysical studies of IgG-receptor interactions, assay development, and receptor characterization.

  • Recombinant extracellular domain expressed in HEK293 cells.
  • C-terminal 6xHis tag for straightforward detection and capture.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Molecular weight approximately 25-30 kDa.
  • Suitable for binding studies, SPR/BLI, and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.