Learn More
Medchemexpress LLC Fc gamma RIIB/CD32b 5ug | 5ug
SDP
Catalog No. 5000186106
Encompass_Preferred
Fc RIIB/CD32b proteins exhibit low affinity as complexes or receptors that aggregate the immunoglobulin Fc region ITIM phosphorylated by Fc RIIB/CD32b recruits inositol phosphatases SHIP1 and SHIP2 inhibits Ras activation down-regulates MAPK activity reduces PLC- function and results in PKC activation Inhibition of MAP kinase pathway and anti-apoptotic kinase Akt inhibit cell proliferation and survival Fc gamma RIIB/CD32b Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIIB/CD32b protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86106 Supplier Medchemexpress LLC Supplier No. HYP768005UG
Fc RIIB/CD32b proteins exhibit low affinity as complexes or receptors that aggregate the immunoglobulin Fc region ITIM phosphorylated by Fc RIIB/CD32b recruits inositol phosphatases SHIP1 and SHIP2 inhibits Ras activation down-regulates MAPK activity reduces PLC- function and results in PKC activation Inhibition of MAP kinase pathway and anti-apoptotic kinase Akt inhibit cell proliferation and survival Fc gamma RIIB/CD32b Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIIB/CD32b protein expressed by HEK293 with C-His labeled tag