Learn More
Medchemexpress LLC Fc gamma RIII/CD16 10ug | 10ug
SDP
Catalog No. 5000187050
Encompass_Preferred
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIII/CD16 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87050 Supplier Medchemexpress LLC Supplier No. HYP7766310UG
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIII/CD16 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag