Learn More
Medchemexpress LLC Fc gamma RIII/CD16 500ug | 500ug
SDP
Catalog No. 5000187452
Encompass_Preferred
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIII/CD16 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87452 Supplier Medchemexpress LLC Supplier No. HYP77663500UG
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIII/CD16 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag