Learn More
Medchemexpress LLC Fc gamma RIII/CD16 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7766350UG
Encompass_Preferred
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIII/CD16 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87282