Learn More
Medchemexpress LLC Fc gamma RIII/CD16 50ug | 50ug
SDP
Catalog No. 5000187233
Encompass_Preferred
The Fc gamma RIII/CD16 protein is a receptor for IgG and is optimally activated upon binding to aggregated antigen-IgG complexes initiating antibody-dependent cellular cytotoxicity (ADCC) and preventing inappropriate effector cell activation It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIII/CD16 Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87233 Supplier Medchemexpress LLC Supplier No. HYP7731450UG
The Fc gamma RIII/CD16 protein is a receptor for IgG and is optimally activated upon binding to aggregated antigen-IgG complexes initiating antibody-dependent cellular cytotoxicity (ADCC) and preventing inappropriate effector cell activation It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIII/CD16 Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived Fc gamma RIII/CD16 protein expressed by HEK293 with C-His labeled tag