Learn More
Genscript Corporation Fc gamma RIII/CD16Biotin Hi
Supplier: Genscript Corporation Z04655500
Immunoglobulin G (IgG) Fc receptors play a critical role in linking IgG antibody-mediated immune responses with cellular effector functions A high resolution map of the binding site on human IgG1 for human Fc gamma RI Fc gamma RIIA Fc gamma RIIB Fc gamma RIIIA and FcRn receptors has been determined A common set of IgG1 residues is involved in binding to all Fc gamma R Fc gamma RII and Fc gamma RIII also utilize residues outside this common set