Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a 100ug
SDP
Catalog No. 5000184440
Encompass_Preferred
Recombinant human FcγRIIIa/CD16a extracellular domain produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized powder formulated from PBS or 20 mM phosphate buffer with 150 mM NaCl, the protein is intended for in vitro binding studies, antibody-dependent cellular cytotoxicity (ADCC) assays, and other applications requiring human Fc receptor protein.
  • Expression in HEK293 cells preserves native glycosylation.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Lyophilized format improves stability and storage life.
  • Suitable for binding assays and ADCC functional studies.
  • Molecular weight consistent with extracellular domain (36-44 kDa).
Catalog No. 50-001-84440 Supplier Medchemexpress LLC Supplier No. HYP70705100UG
Recombinant human FcγRIIIa/CD16a extracellular domain produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized powder formulated from PBS or 20 mM phosphate buffer with 150 mM NaCl, the protein is intended for in vitro binding studies, antibody-dependent cellular cytotoxicity (ADCC) assays, and other applications requiring human Fc receptor protein.
  • Expression in HEK293 cells preserves native glycosylation.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Lyophilized format improves stability and storage life.
  • Suitable for binding assays and ADCC functional studies.
  • Molecular weight consistent with extracellular domain (36-44 kDa).