Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a 10ug | 10ug
SDP
Catalog No. 5000194152
Encompass_Preferred
Fc gamma RIIIA/CD16a protein is a receptor for the Fc fragment of IgG and activates antibody-dependent cellular cytotoxicity (ADCC) upon binding to antigen-IgG complexes It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIIIA/CD16a Protein Human (F176V HEK293 SUMO-His) is the recombinant human-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-His N-SUMO labeled tag and F176V mutation
Catalog No. 50-001-94152 Supplier Medchemexpress LLC Supplier No. HYP7867610UG
Fc gamma RIIIA/CD16a protein is a receptor for the Fc fragment of IgG and activates antibody-dependent cellular cytotoxicity (ADCC) upon binding to antigen-IgG complexes It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIIIA/CD16a Protein Human (F176V HEK293 SUMO-His) is the recombinant human-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-His N-SUMO labeled tag and F176V mutation