Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems CHO/Human CD16a (FcγRIIIa) 158V Stable Cell Line (High Expression), 2 vials

Catalog No. 502357309
Encompass_Preferred

This CHO-derived stable cell line expresses full-length human CD16a (FcγRIIIa) carrying the 158V variant at high levels. Supplied frozen in cryopreservation medium as two vials (1 mL per vial), the cell line is intended for cell-based assays that require CD16a-dependent binding or antibody crosslinking, for example co-culture with 4-1BB reporter cells to evaluate agonist activity. Culture and selection conditions are provided in the product datasheet.

  • High expression of full-length human CD16a (158V) for robust assay readouts.
  • Adherent CHO host suitable for standard cell culture workflows.
  • Supplied frozen with at least 5x10^6 cells per 1 mL vial.
  • Hygromycin B selection marker for stable maintenance (20 μg/mL).
  • QC tested: mycoplasma negative and sterility tested.
  • Optimized growth conditions provided (F-12K + 10% FBS).

Catalog No. 50-235-7309 Supplier ACROBiosystems Supplier No. SCCHOATP059H2VIALS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This CHO-derived stable cell line expresses full-length human CD16a (FcγRIIIa) carrying the 158V variant at high levels. Supplied frozen in cryopreservation medium as two vials (1 mL per vial), the cell line is intended for cell-based assays that require CD16a-dependent binding or antibody crosslinking, for example co-culture with 4-1BB reporter cells to evaluate agonist activity. Culture and selection conditions are provided in the product datasheet.

  • High expression of full-length human CD16a (158V) for robust assay readouts.
  • Adherent CHO host suitable for standard cell culture workflows.
  • Supplied frozen with at least 5x10^6 cells per 1 mL vial.
  • Hygromycin B selection marker for stable maintenance (20 μg/mL).
  • QC tested: mycoplasma negative and sterility tested.
  • Optimized growth conditions provided (F-12K + 10% FBS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.