Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a 50ug | 50ug
SDP
Catalog No. 5000187523
Encompass_Preferred
Fc gamma RIIIA/CD16a protein is a receptor for the Fc fragment of IgG and activates antibody-dependent cellular cytotoxicity (ADCC) upon binding to antigen-IgG complexes It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIIIA/CD16a Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87523 Supplier Medchemexpress LLC Supplier No. HYP7809950UG
Fc gamma RIIIA/CD16a protein is a receptor for the Fc fragment of IgG and activates antibody-dependent cellular cytotoxicity (ADCC) upon binding to antigen-IgG complexes It mediates IgG effector functions on NK cells generating memory-like adaptive NK cells to efficiently eliminate virally infected cells Fc gamma RIIIA/CD16a Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-Avi C-His labeled tag