Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a 5ug | 5ug
SDP
Catalog No. 5000195135
Encompass_Preferred
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIIIA/CD16a Protein Cynomolgus (Biotinylated HEK293 His-Avi) is the recombinant cynomolgus-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-95135 Supplier Medchemexpress LLC Supplier No. HYP788795UG
The Fc RIIIA/CD16a protein is a receptor for IgG and is optimally activated upon binding of aggregated antigen-IgG complexes inducing antibody-dependent cellular cytotoxicity (ADCC) It mediates IgG effector function on natural killer cells limiting viral load Fc gamma RIIIA/CD16a Protein Cynomolgus (Biotinylated HEK293 His-Avi) is the recombinant cynomolgus-derived Fc gamma RIIIA/CD16a protein expressed by HEK293 with C-Avi C-His labeled tag