Learn More
Genscript Corporation Fc gamma RIIIA/CD16a F176Bi
Catalog No. 5000058191
Encompass
Human Fc gamma RIIIA/CD16a Protein is a receptor for the Fc region of IgG Binds complexed or aggregated IgG and also monomeric IgG Mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses such as phagocytosis
Catalog No. 50-000-58191 Supplier Genscript Corporation Supplier No. Z04082500
Human Fc gamma RIIIA/CD16a Protein is a receptor for the Fc region of IgG Binds complexed or aggregated IgG and also monomeric IgG Mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses such as phagocytosis