Learn More
Medchemexpress LLC Fc gamma RIIIA (CD16a) protein | 98.0% | 50 UG
SDP
Catalog No. 5000292517
Encompass_Preferred
Recombinant rat Fc gamma RIIIA (CD16a) protein expressed in HEK293 cells with a C-terminal His tag, supplied as a purified, soluble preparation (supplier indicates 50 μg). The protein is intended for use in binding assays, receptor characterization, and studies of antibody-dependent cellular cytotoxicity (ADCC).
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Expressed in mammalian cells for native glycosylation.
  • C-terminal His tag facilitates immobilization and detection.
  • Recommended reconstitution concentration ≥100 μg/mL.
Catalog No. 50-002-92517 Supplier Medchemexpress LLC Supplier No. HYP7562150UG
Recombinant rat Fc gamma RIIIA (CD16a) protein expressed in HEK293 cells with a C-terminal His tag, supplied as a purified, soluble preparation (supplier indicates 50 μg). The protein is intended for use in binding assays, receptor characterization, and studies of antibody-dependent cellular cytotoxicity (ADCC).
  • Purity ≥98.0% by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Expressed in mammalian cells for native glycosylation.
  • C-terminal His tag facilitates immobilization and detection.
  • Recommended reconstitution concentration ≥100 μg/mL.