Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a protein, rat | 5 UG
SDP
Catalog No. 5000292622
Encompass_Preferred
Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain expressed in HEK293 cells with a C-terminal His tag. Supplied as a small, purified protein for receptor binding studies, antibody-dependent cellular cytotoxicity (ADCC) assays, and in vitro functional characterization.
  • Rat extracellular domain expressed in HEK293 cells.
  • C-terminal His tag for convenient detection and purification.
  • Glycosylated to retain native binding properties.
  • Suitable for binding assays and receptor characterization.
  • Shipped frozen with recommended storage for long-term stability.
Catalog No. 50-002-92622 Supplier Medchemexpress LLC Supplier No. HYP75621A5UG
Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain expressed in HEK293 cells with a C-terminal His tag. Supplied as a small, purified protein for receptor binding studies, antibody-dependent cellular cytotoxicity (ADCC) assays, and in vitro functional characterization.
  • Rat extracellular domain expressed in HEK293 cells.
  • C-terminal His tag for convenient detection and purification.
  • Glycosylated to retain native binding properties.
  • Suitable for binding assays and receptor characterization.
  • Shipped frozen with recommended storage for long-term stability.