Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fc gamma RIIIA/CD16a protein, rat | >95.0% | 10 UG
SDP

Catalog No. 5000290593
Encompass_Preferred

Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain expressed in HEK293 cells, purified and supplied as a 0.22 μm-filtered PBS solution with a C-terminal His tag. Intended for in vitro Fc receptor binding studies, ADCC-related assays, and biophysical characterization.

  • Expression system: HEK293 (mammalian)
  • Tag: C-terminal His tag
  • Format: 0.22 μm-filtered solution in PBS, pH 7.4
  • Molecular weight: 28-33 kDa
  • Purity: >95% by reducing SDS-PAGE
  • Available sizes: microgram quantities (5, 10, 50, 100 μg)
  • Applications: Fc receptor binding, ADCC assays, and biophysical studies

Catalog No. 50-002-90593 Supplier Medchemexpress LLC Supplier No. HYP75621A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain expressed in HEK293 cells, purified and supplied as a 0.22 μm-filtered PBS solution with a C-terminal His tag. Intended for in vitro Fc receptor binding studies, ADCC-related assays, and biophysical characterization.

  • Expression system: HEK293 (mammalian)
  • Tag: C-terminal His tag
  • Format: 0.22 μm-filtered solution in PBS, pH 7.4
  • Molecular weight: 28-33 kDa
  • Purity: >95% by reducing SDS-PAGE
  • Available sizes: microgram quantities (5, 10, 50, 100 μg)
  • Applications: Fc receptor binding, ADCC assays, and biophysical studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.