Learn More
Medchemexpress LLC Fc gamma RIIIA (CD16a) protein, rat | ≥98.0% | 10 UG
SDP
Catalog No. 5000292532
Encompass_Preferred
Recombinant rat Fc gamma RIIIA (CD16a) protein expressed in HEK293 cells with a C-terminal His tag, supplied as a solution. Intended for receptor-binding studies and immune-effector function assays (for example, ADCC), the product is characterized by lot documentation suitable for biochemical and assay applications.
  • Recombinant rat sequence expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Provided as a liquid solution for immediate use.
  • High purity, ≥98.0% as determined by SDS-PAGE.
  • Suitable for receptor binding and ADCC functional assays.
Catalog No. 50-002-92532 Supplier Medchemexpress LLC Supplier No. HYP7562110UG
Recombinant rat Fc gamma RIIIA (CD16a) protein expressed in HEK293 cells with a C-terminal His tag, supplied as a solution. Intended for receptor-binding studies and immune-effector function assays (for example, ADCC), the product is characterized by lot documentation suitable for biochemical and assay applications.
  • Recombinant rat sequence expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Provided as a liquid solution for immediate use.
  • High purity, ≥98.0% as determined by SDS-PAGE.
  • Suitable for receptor binding and ADCC functional assays.