Learn More
Medchemexpress LLC Fc gamma RIIIA (CD16a) protein, rat | ≥98.0% | 5 UG
SDP
Catalog No. 5000292551
Encompass_Preferred
Recombinant rat Fc gamma receptor IIIA (FcγRIIIA/CD16a) produced in HEK293 cells with a C-terminal His tag and supplied in solution. The protein is intended for in vitro studies of Fc-IgG interactions, antibody-dependent cellular cytotoxicity (ADCC), and receptor characterization; available in small quantities (5 μg) for assay-scale experiments.
  • Recombinant rat FcγRIIIA/CD16a produced in mammalian HEK293 cells.
  • C-terminal His tag enables purification and detection.
  • Supplied in solution for immediate use in biochemical and cell-based assays.
  • Suitable for Fc-IgG binding studies and ADCC assay development.
  • High purity verified by reducing SDS-PAGE (batch result ≥98%).
Catalog No. 50-002-92551 Supplier Medchemexpress LLC Supplier No. HYP756215UG
Recombinant rat Fc gamma receptor IIIA (FcγRIIIA/CD16a) produced in HEK293 cells with a C-terminal His tag and supplied in solution. The protein is intended for in vitro studies of Fc-IgG interactions, antibody-dependent cellular cytotoxicity (ADCC), and receptor characterization; available in small quantities (5 μg) for assay-scale experiments.
  • Recombinant rat FcγRIIIA/CD16a produced in mammalian HEK293 cells.
  • C-terminal His tag enables purification and detection.
  • Supplied in solution for immediate use in biochemical and cell-based assays.
  • Suitable for Fc-IgG binding studies and ADCC assay development.
  • High purity verified by reducing SDS-PAGE (batch result ≥98%).