Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fc gamma RIIIA/CD16a protein, rat (HEK293, C-His) | ≥98.0% | 100 UG
SDP

Catalog No. 5000292485
Encompass_Preferred

Recombinant rat Fc gamma RIIIA/CD16a protein expressed in HEK293 cells and supplied with a C-terminal His tag for detection and purification. The material is provided lyophilized with accompanying COA and datasheet and is intended for use in binding and functional assays, including ADCC-related studies.

  • Recombinant rat FcγRIIIA/CD16a expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity, ≥98.0% (COA) and >95% by reducing SDS-PAGE.
  • Suitable for binding and ADCC-related functional assays; ED50 ≈93 ng/mL for human IgG1 binding.
  • Lyophilized from PBS, pH 7.4; reconstitute to ≥100 μg/mL.
  • Store at -20°C for long-term storage; 4°C stability for 1 week after reconstitution.
  • Carrier protein recommended for reconstitution/storage (0.1% BSA, 5% HSA, 10% FBS, or 5% trehalose).

Catalog No. 50-002-92485 Supplier Medchemexpress LLC Supplier No. HYP75621A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Fc gamma RIIIA/CD16a protein expressed in HEK293 cells and supplied with a C-terminal His tag for detection and purification. The material is provided lyophilized with accompanying COA and datasheet and is intended for use in binding and functional assays, including ADCC-related studies.

  • Recombinant rat FcγRIIIA/CD16a expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity, ≥98.0% (COA) and >95% by reducing SDS-PAGE.
  • Suitable for binding and ADCC-related functional assays; ED50 ≈93 ng/mL for human IgG1 binding.
  • Lyophilized from PBS, pH 7.4; reconstitute to ≥100 μg/mL.
  • Store at -20°C for long-term storage; 4°C stability for 1 week after reconstitution.
  • Carrier protein recommended for reconstitution/storage (0.1% BSA, 5% HSA, 10% FBS, or 5% trehalose).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.