Learn More
Medchemexpress LLC Fc gamma RIIIA/CD16a protein, rat (HEK293, His) | ≥98.0% | 500 UG
SDP
Catalog No. 5000290454
Encompass_Preferred
Recombinant rat Fc gamma RIIIA/CD16a protein produced in HEK293 cells with a C-terminal His tag. The protein binds the Fc region of IgG and is suitable for antibody-binding studies, ADCC-related assays, and assay development. Supplied lyophilized with recommended reconstitution and storage instructions.
  • Recombinant rat Fc gamma RIIIA/CD16a ectodomain expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • High purity (≥98.0% by reducing SDS-PAGE) for consistent performance.
  • Lyophilized formulation from PBS for improved stability.
  • Recommended reconstitution ≥100 μg/mL and aliquot storage at -20°C or -80°C.
  • Suitable for binding assays, ADCC studies, and antibody interaction analyses.
Catalog No. 50-002-90454 Supplier Medchemexpress LLC Supplier No. HYP75621A500UG
Recombinant rat Fc gamma RIIIA/CD16a protein produced in HEK293 cells with a C-terminal His tag. The protein binds the Fc region of IgG and is suitable for antibody-binding studies, ADCC-related assays, and assay development. Supplied lyophilized with recommended reconstitution and storage instructions.
  • Recombinant rat Fc gamma RIIIA/CD16a ectodomain expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • High purity (≥98.0% by reducing SDS-PAGE) for consistent performance.
  • Lyophilized formulation from PBS for improved stability.
  • Recommended reconstitution ≥100 μg/mL and aliquot storage at -20°C or -80°C.
  • Suitable for binding assays, ADCC studies, and antibody interaction analyses.