Learn More
Medchemexpress LLC Fc gamma RIIIA (CD16a) recombinant protein, rat | >95.0% | 1 MG
SDP
Catalog No. 5000292518
Encompass_Preferred
Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain produced in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution for biochemical and functional studies of Fc receptor binding and antibody-dependent cellular cytotoxicity (ADCC). Multiple pack sizes are available, including a 1 mg solution.
  • Produced in HEK293 cells to support native-like glycosylation.
  • C-terminal His tag for straightforward purification and detection.
  • Supplied as a 0.22 μm filtered solution for immediate use.
  • High purity (>95.0%) as tested by SDS-PAGE/HPLC.
  • Suitable for binding assays, ADCC functional studies, and antibody characterization.
Catalog No. 50-002-92518 Supplier Medchemexpress LLC Supplier No. HYP75621A1MG
Recombinant rat Fc gamma RIIIA (CD16a) extracellular domain produced in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution for biochemical and functional studies of Fc receptor binding and antibody-dependent cellular cytotoxicity (ADCC). Multiple pack sizes are available, including a 1 mg solution.
  • Produced in HEK293 cells to support native-like glycosylation.
  • C-terminal His tag for straightforward purification and detection.
  • Supplied as a 0.22 μm filtered solution for immediate use.
  • High purity (>95.0%) as tested by SDS-PAGE/HPLC.
  • Suitable for binding assays, ADCC functional studies, and antibody characterization.