Learn More
Genscript Corporation Fc gamma RIIIA/CD16a V176Bi
Catalog No. 5000058796
Encompass
The Human Fc gamma RIIIA/CD16a Protein is a receptor for the Fc region of IgG Binds complexed or aggregated IgG and also monomeric IgG Mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses such as phagocytosis
Catalog No. 50-000-58796 Supplier Genscript Corporation Supplier No. Z04451500
The Human Fc gamma RIIIA/CD16a Protein is a receptor for the Fc region of IgG Binds complexed or aggregated IgG and also monomeric IgG Mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses such as phagocytosis