Learn More
Medchemexpress LLC Fc gamma RIIIB/CD16b 50ug | 50ug
SDP
Catalog No. 5000187705
Encompass_Preferred
Fc gamma RIIIB/CD16b Protein a low-affinity receptor for IgG binds complexed or monomeric IgG Unlike Fc gamma RIIIA it cannot mediate antibody-dependent cytotoxicity or phagocytosis Instead Fc gamma RIIIB may act as a trap for immune complexes circulating without activating neutrophils Existing as a monomer it interacts with INPP5D/SHIP1 implying its role in intracellular signaling pathways linked to immune responses Fc gamma RIIIB/CD16b Protein Human (Biotinylated NA2 HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIIB/CD16b protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87705 Supplier Medchemexpress LLC Supplier No. HYP7812250UG
Fc gamma RIIIB/CD16b Protein a low-affinity receptor for IgG binds complexed or monomeric IgG Unlike Fc gamma RIIIA it cannot mediate antibody-dependent cytotoxicity or phagocytosis Instead Fc gamma RIIIB may act as a trap for immune complexes circulating without activating neutrophils Existing as a monomer it interacts with INPP5D/SHIP1 implying its role in intracellular signaling pathways linked to immune responses Fc gamma RIIIB/CD16b Protein Human (Biotinylated NA2 HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIIB/CD16b protein expressed by HEK293 with C-Avi C-His labeled tag