Learn More
Medchemexpress LLC Fc gamma RIIIB (CD16b) protein, human, biotinylated | 100UG
SDP
Catalog No. 5000293055
Encompass_Preferred
Fc gamma RIIIB/CD16b Protein a low-affinity receptor for IgG binds complexed or monomeric IgG Unlike Fc gamma RIIIA it cannot mediate antibody-dependent cytotoxicity or phagocytosis Instead Fc gamma RIIIB may act as a trap for immune complexes circulating without activating neutrophils Existing as a monomer it interacts with INPP5D/SHIP1 implying its role in intracellular signaling pathways linked to immune responses Fc gamma RIIIB/CD16b Protein Human (Biotinylated NA2) is the recombinant human-derived Fc gamma RIIIB/CD16b protein expressed by E coli with tag free
Catalog No. 50-002-93055 Supplier Medchemexpress LLC Supplier No. HYP75204100UG
Fc gamma RIIIB/CD16b Protein a low-affinity receptor for IgG binds complexed or monomeric IgG Unlike Fc gamma RIIIA it cannot mediate antibody-dependent cytotoxicity or phagocytosis Instead Fc gamma RIIIB may act as a trap for immune complexes circulating without activating neutrophils Existing as a monomer it interacts with INPP5D/SHIP1 implying its role in intracellular signaling pathways linked to immune responses Fc gamma RIIIB/CD16b Protein Human (Biotinylated NA2) is the recombinant human-derived Fc gamma RIIIB/CD16b protein expressed by E coli with tag free