Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fc gamma RIIIB/CD16b protein, human (biotinylated, NA2) | >90.0% | 20 UG
SDP

Catalog No. 5000292689
Encompass_Preferred

Recombinant human Fc gamma RIIIB (CD16b) protein, biotinylated (NA2), supplied as a tag-free, ~20 kDa receptor fragment for use in biochemical and binding assays that involve IgG interactions.

  • Biotinylated conjugation for affinity detection.
  • Recombinant human sequence expressed in E. coli.
  • Tag free preparation to avoid epitope interference.
  • Approximately 20 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in small research pack suitable for assay development.
  • Stable when stored at -20 °C; freeze aliquots for long-term storage.
  • Suitable for binding and interaction studies with IgG.

Catalog No. 50-002-92689 Supplier Medchemexpress LLC Supplier No. HYP7520420UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Fc gamma RIIIB (CD16b) protein, biotinylated (NA2), supplied as a tag-free, ~20 kDa receptor fragment for use in biochemical and binding assays that involve IgG interactions.

  • Biotinylated conjugation for affinity detection.
  • Recombinant human sequence expressed in E. coli.
  • Tag free preparation to avoid epitope interference.
  • Approximately 20 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Supplied in small research pack suitable for assay development.
  • Stable when stored at -20 °C; freeze aliquots for long-term storage.
  • Suitable for binding and interaction studies with IgG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.