Learn More
Enzo Life Sciences Fc (human) : RANKL (soluble) (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522131C010
Encompass_Preferred
Alternative name: TNFSF 11, CD254, TRANCE, ODF, OPGL. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse RANKL (aa 157-316) is fused to the Fc portion of human IgG1. Specificity: Binds to mouse RANK. Long Term Storage: -20°C.
Catalog No. 50-200-9445