Learn More
AdipoGen Fc hRANKL Soluble ms
SDP
Catalog No. 502562929
Encompass_Preferred
Protein. Mouse RANKL aa 157-316 is fused at the N-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds to mouse RANK. Purity >95% SDS-PAGE. RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T cell proliferation. Important regulator of interactions between T cells and dendritic cells and may play a role in the regulation of the T cell-dependent immune response. RANKL plays an important role in the progression of Breast cancer.
Catalog No. 50-256-2929 Supplier AdipoGen Supplier No. AG40B0059C010
Protein. Mouse RANKL aa 157-316 is fused at the N-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds to mouse RANK. Purity >95% SDS-PAGE. RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T cell proliferation. Important regulator of interactions between T cells and dendritic cells and may play a role in the regulation of the T cell-dependent immune response. RANKL plays an important role in the progression of Breast cancer.