Learn More
AdipoGen Fc (human):Dectin-1 (mouse) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0138C050
Encompass_Preferred
Fc (human):Dectin-1 (mouse) (rec.) (50 µg), AdipoGen Life Sciences. Protein. Mouse dectin-1 (aa 72-244) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Dectin-1 and dectin-2 are type II transmembrane proteins of the C-type lectin family. They contain single carbohydrate recognition domains (CRDs) in their extracellular region. Dectin-1 and dectin-2 are expressed in macrophages and dendritic cells. Dectin-1 recognizes beta-glucans with its CRD and signals through its immunoreceptor tyrosine-based activation motif (ITAM)-like motif in the cytoplasmic domain, whereas dectin-2 recognizes alpha-mannans and transduces its signal through association with the ITAM-containing Fc receptor gamma-chain.
Catalog No. 50-148-9426