Learn More
AdipoGen Fc (human):FasL, Soluble (human) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40B0132C010
Encompass_Preferred
Protein. Human FasL (aa 139-281) is fused at the N-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse Fas. Purity: >95% (SDS-PAGE). Fc (human):FasL, Soluble (human) is a high activity construct in which two trimeric FasL are artificially linked via the Fc binding domain of human IgG1. This construct very effectively simulates the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Catalog No. 50-205-3212