Learn More
AdipoGen Fc (human):FLT3 Ligand (human) (rec.) (100 µg)
SDP
Supplier:  AdipoGen AG40B0119C100
Encompass_Preferred
Protein. The receptor-binding domain of human FLT3L (aa1-185) is fused at the N-terminus to the Fc portion of human IgG1. Source: CHO cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Liquid. In PBS (sterile). Binds to human and mouse FLT3 receptor. Purity: >95% (SDS-PAGE). FMS-like tyrosine kinase 3 ligand (FLT3L) acts as a growth factor that increases the number of immune cells (lymphocytes (B cells and T cells)) by activating the hematopoietic progenitors. It binds to FLT3 (CD135) which is found on multipotent progenitor (MPP) and common lymphoid progenitor (CLP) cells in mice. FLT3L induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3L is crucial for steady-state pDC (plasmacytoid dendritic cells) and cDC (classical dendritic cells) development. Deficiency of FLT3L causes a dramatic decrease in DC numbers, whereas increasing its availability increase in DC numbers.
Catalog No. 50-205-3192