Learn More
AdipoGen Fc (human) IgG1 Control (rec.) (1 mg)
SDP
Supplier:  AdipoGen AG35B0007M001
Encompass_Preferred
Protein. Recombinant Fc portion of human IgG1 (aa 105-330). Source: CHO cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Negative control in experiments with recombinant proteins fused to human IgG1 Fc.
Catalog No. 50-205-3057