Learn More
AdipoGen Fc (human):OX40L, Soluble (human) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01723010
Encompass_Preferred
Protein. The extracellular domain of human OX40L (aa 50-183) is fused at the N-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). OX40L [CD252; TNFSF4] is a membrane-expressed cytokine that belongs to the TNF ligand family. It acts as costimulator through interaction with its receptor OX40 on T cells, stimulating T cell activation, proliferation and cytokine production. OX40L is expressed on antigen presenting cells including B cells, dendritic cells, mast cells and endothelium. Fc (human):OX40L, Soluble (human) (rec.) is a high activity construct in which two trimeric OX40L ligands are artificially linked via the Fc domain of human immunoglobulin.
Catalog No. 50-205-3269