Learn More
AdipoGen Fc human IgG1 Ctrl rec
SDP
Supplier:  AdipoGen AG35B00072050
Encompass_Preferred
Protein. Recombinant Fc portion of human IgG1 aa 105-330. Lyophilized. Contains PBS. Negative control in experiments with recombinant proteins fused to human IgG1 Fc.
Catalog No. 50-256-3085