Learn More
AdipoGen Fc human IgG2 Ctrl
SDP
Supplier:  AdipoGen AG35B00092050
Encompass_Preferred
Protein. Recombinant Fc portion of human IgG2b. Lyophilized. Contains PBS. Fc human IgG2 Control rec. AG-35B-0009 serves as a negative control in experiments with monoclonal antibodies recombinant AG-27B-XXXX containing a Fc human IgG2.
Catalog No. 50-256-3082