Learn More
AdipoGen Fc humanFNDC4 rec
SDP
Catalog No. 502562867
Encompass_Preferred
Recombinant Protein. FNDC4 extracellular domain, human aa 45-167 / mouse aa 41-163 is fused at the N-terminus to the Fc portion of human IgG1. FNDC4 extracellular domain has 100% identity between human, mouse, rat, dog and monkey. Lyophilized. Contains PBS. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone irisin. Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure. FNDC4 is an ortholog of FNDC5 with 50% identity and 86% similarity compared to Irisin. FNDC4 as well as FNDC5 are extremely well conserved between species. The human FNDC4 gene is highly enriched in liver, brain tissue and adipocytes.
Catalog No. 50-256-2867 Supplier AdipoGen Supplier No. AG40B0213C050
Recombinant Protein. FNDC4 extracellular domain, human aa 45-167 / mouse aa 41-163 is fused at the N-terminus to the Fc portion of human IgG1. FNDC4 extracellular domain has 100% identity between human, mouse, rat, dog and monkey. Lyophilized. Contains PBS. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone irisin. Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure. FNDC4 is an ortholog of FNDC5 with 50% identity and 86% similarity compared to Irisin. FNDC4 as well as FNDC5 are extremely well conserved between species. The human FNDC4 gene is highly enriched in liver, brain tissue and adipocytes.