Learn More
AdipoGen Fc mouse IgG2b Ctrl
SDP
Supplier:  AdipoGen AG35B0008C050
Encompass_Preferred
Protein. Recombinant Fc portion of mouse IgG2b. Lyophilized. Contains PBS. Fc mouse IgG2b Control rec. Prod. No. AG-35B-0008 serves as a negative control in experiments with anti-APRIL mouse, mAb rec. Blocking APRY-1-1 Prod. No. AG-27B-0001.
Catalog No. 50-256-3083