Learn More
AdipoGen Fc ms IgG2a Ctrl recn-l
SDP
Supplier:  AdipoGen AG35B00173010
Encompass_Preferred
Recombinant Protein. Fc mouse IgG2a 100-330 including 4 mutations. Source W2. Lyophilized. Contains PBS. Negative control in experiments with recombinant proteins fused to Fc mouse IgG2a non-lytic e.g. Prod. No. AG-40B-0231. This protein is not an appropriate negative control for other standard Fc protein constructs. Mutations to the complement C1q and FcgR I binding sites of the IgGs Fc fragment render the non-lytic proteins incapable of antibody-directed cytotoxicity ADCC and complement-directed cytotoxicity CDC.
Catalog No. 50-256-3069