Learn More
INNOVEX BIOSCIENCES UNIVERSAL Fc RECEPTORS BLOCKER NON-ANTIBODY BASED 15 ML
SDP
Catalog No. 50486806
Encompass_Preferred
Innovex peptide-based Fc Blocker universally blocks all types Fc receptors (I, II, III) of human, mouse & all animal species; Contains NO antibody, Ig or Ig fragments, is used for IHC, IF & flow cytometry
Catalog No. 50-486-806 Supplier INNOVEX BIOSCIENCES Supplier No. NB30915
Innovex peptide-based Fc Blocker universally blocks all types Fc receptors (I, II, III) of human, mouse & all animal species; Contains NO antibody, Ig or Ig fragments, is used for IHC, IF & flow cytometry