Learn More
Medchemexpress LLC HY-P1104A 1mg Medchemexpress, FC131 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1104A1MG
Encompass_Preferred
Medchemexpress, HY-P1104A 1mg FC131 (TFA) CAS: FC131 TFA is a CXCR4 antagonist, inhibits [125I]-SDF-1 binding to CXCR4, with an IC50 of 4.5 nM. Anti-HIV activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0525