Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FC131 TFA | 842166-42-1 | 99.9% | 843.85 | 10 MG
SDP

Catalog No. 5000423645
Encompass_Preferred

FC131 TFA is a CXCR4 antagonist that inhibits [125I]-SDF-1 binding to CXCR4 with an IC50 of 4.5 nM. It also exhibits anti-HIV activity. It is for research use only and not sold to patients.

  • Inhibits [125I]-SDF-1 binding to CXCR4 with an IC50 of 4.5 nM
  • Exhibits anti-HIV activity
  • Appearance: solid
  • Color: white to off-white
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-23645 Supplier Medchemexpress LLC Supplier No. HYP1104A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FC131 TFA is a CXCR4 antagonist that inhibits [125I]-SDF-1 binding to CXCR4 with an IC50 of 4.5 nM. It also exhibits anti-HIV activity. It is for research use only and not sold to patients.

  • Inhibits [125I]-SDF-1 binding to CXCR4 with an IC50 of 4.5 nM
  • Exhibits anti-HIV activity
  • Appearance: solid
  • Color: white to off-white
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.