Learn More
Julabo USA Inc Each, FC1600 recirculating chiller 1.6kW
SDP
Catalog No. 507533970
Encompass_Preferred
Chillers, Julabo, FC1600 Recirculating Cooler -20...+80 °C, 1.65kW cooling power; pump 20L/min to 7.3 PSI. JULABO FC units feature two separate Multi-Displays (LED) for actual and set point values. Models designated FCW are water-cooled which reduces the noise level and reduced heat generation to the laboratory environment. Units are conveniently filled from the front and have a removable venting grid.FC units feature a built-in 1.2kW heater to facilitate cooling and heating applications for temperatures from -25 °C to +80 °C with an adjustable ratio for inlet/outlet temperature. Models FC1600T and FCW2500T have a Pt100 sensor connection for external measurement and control, temperature recorder and connections for external programming. All FC units have RS232 interface with connections for standby input and alarm output. FC models supplied with 2 barbed fittings for 8 and 12 mm internal ID tubing. Pump connections M16x1 male. 230V/60Hz.
Catalog No. 50-753-3970 Supplier Julabo USA Inc Supplier No. 9600160.13
Add to Cart
Add to Cart
Request a Quote
Chillers, Julabo, FC1600 Recirculating Cooler -20...+80 °C, 1.65kW cooling power; pump 20L/min to 7.3 PSI. JULABO FC units feature two separate Multi-Displays (LED) for actual and set point values. Models designated FCW are water-cooled which reduces the noise level and reduced heat generation to the laboratory environment. Units are conveniently filled from the front and have a removable venting grid.FC units feature a built-in 1.2kW heater to facilitate cooling and heating applications for temperatures from -25 °C to +80 °C with an adjustable ratio for inlet/outlet temperature. Models FC1600T and FCW2500T have a Pt100 sensor connection for external measurement and control, temperature recorder and connections for external programming. All FC units have RS232 interface with connections for standby input and alarm output. FC models supplied with 2 barbed fittings for 8 and 12 mm internal ID tubing. Pump connections M16x1 male. 230V/60Hz.