Learn More
Ohaus FC5706P Multi Centrifuge
Catalog No. 01259090 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Benchtop centrifuge
Catalog No. Product Type
01-259-090 Benchtop centrifuge
1 options
Catalog No. 01-259-090 Supplier Ohaus Supplier No. 31060778
Add to Cart
Add to Cart
Frontier FC5706P offers 5 rotor options, including fixed-angle (18 x 1.5/2.0 mL) and swing-out (4 x 10 mL) configurations.
  • Simple and Intuitive Operation: Convenient controls with an intuitive interface allow quick and responsive handling, even while wearing gloves.
  • Versatile Tube Compatibility: Accommodates standard 50 mL and 15 mL tubes, including common blood and urine tubes. With accessories, rotors can also be adapted for smaller sizes down to 1.5 mL.
  • Space-Saving Design: Compact construction integrates all essential functionalities within a small footprint, saving valuable bench space in the lab.
  • Flexible Rotor Options: Offers 5 rotor choices including fixed-angle (18 x 1.5/2.0 mL tubes) and swing-out (4 x 10 mL blood/urine tubes). Rotors are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 6 x 50 mL
Max. RCF 4,427 x g
Max. Speed 6,000 rpm
Refrigerated No
Product Type Benchtop centrifuge